Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-[1,2-Ethenediylbis(4,1-phenyleneiminocarbonyl)]bis(N-butyl-N,N-dimethylbenzenemethanaminium) Dichloride 94.0+%, TCI America™
SDP

Catalog No. E11271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11271G Supplier TCI America Supplier No. E11271G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1296211-78-3
Molecular Formula C42H52Cl2N4O2
Molecular Weight (g/mol) 715.804
InChI Key HHTANURNIWSYAS-UHFFFAOYSA-N
PubChem CID 131637023
IUPAC Name butyl-[[4-[[4-[2-[4-[[4-[[butyl(dimethyl)azaniumyl]methyl]benzoyl]amino]phenyl]ethynyl]phenyl]carbamoyl]phenyl]methyl]-dimethylazanium;dichloride
SMILES CCCC[N+](C)(C)CC1=CC=C(C=C1)C(=O)NC2=CC=C(C=C2)C#CC3=CC=C(C=C3)NC(=O)C4=CC=C(C=C4)C[N+](C)(C)CCCC.[Cl-].[Cl-]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 717.82
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-[1,2-Ethenediylbis(4,1-phenyleneiminocarbonyl)]bis(N-butyl-N,N-dimethylbenzenemethanamini
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.