Learn More
Medchemexpress LLC 4-[4-(1,3-benzodioxol-5-yl)-5-pyridin-2-yl-1H-imidazol-2-yl]benzamide | 301836-41-9 | MFCD05865244 | 99.4% | 384.39 g/mol | C22H16N4O3 | 1 MG

Supplier: Medchemexpress LLC HY10431R1MG
SB-431542 (Standard) is an analytical standard of a TGF-β receptor kinase inhibitor used as a reference material for qualitative and quantitative methods, commonly applied in HPLC, GC, and MS, and in research of TGF-β signaling and cancer-related pathways.
- Analytical standard for HPLC, GC, and MS.
- High purity: 99.4% (rounded).
- Molecular formula: C22H16N4O3.
- Molecular weight: 384.39 g/mol.
- Physical form: solid suitable for analytical preparation.
- Common applications: signal transduction and cancer research.
- Typical package size: 1 mg available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.