Learn More
Medchemexpress LLC 4-[4-(1,3-benzodioxol-5-yl)-5-pyridin-2-yl-1H-imidazol-2-yl]benzamide | 301836-41-9 | MFCD05865244; MFCD11045982 | 99.4% | 384.39 g·mol⁻¹ | C22H16N4O3 | 10 MG
SDP
Catalog No. 5000242195
Encompass_Preferred
SB-431542 (Standard) is an analytical reference of a small-molecule TGF-β type I receptor inhibitor used in biochemical and cell signaling research. It is provided as a 10 mg standard for assay development, method validation, and pharmacology studies, with purity and batch information documented in the Certificate of Analysis.
Catalog No. 50-002-42195 Supplier Medchemexpress LLC Supplier No. HY10431R10MG
SB-431542 (Standard) is an analytical reference of a small-molecule TGF-β type I receptor inhibitor used in biochemical and cell signaling research. It is provided as a 10 mg standard for assay development, method validation, and pharmacology studies, with purity and batch information documented in the Certificate of Analysis.