Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-(2,3-Dihydrothieno[3,4-b][1,4]dioxine-5,7-diyl)bis[N,N-bis(4-methoxyphenyl)aniline] 98.0+%, TCI America™
SDP

Catalog No. D5155200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5155200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D5155200MG Supplier TCI America Supplier No. D5155200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1622008-73-4
Molecular Formula C46H40N2O6S
Molecular Weight (g/mol) 748.89
MDL Number MFCD28369275
InChI Key JOJPNOVYPGLCMM-UHFFFAOYSA-N
Synonym H101
PubChem CID 102130343
IUPAC Name 4-(7-{4-[bis(4-methoxyphenyl)amino]phenyl}-2H,3H-thieno[3,4-b][1,4]dioxin-5-yl)-N,N-bis(4-methoxyphenyl)aniline
SMILES COC1=CC=C(C=C1)N(C1=CC=C(OC)C=C1)C1=CC=C(C=C1)C1=C2OCCOC2=C(S1)C1=CC=C(C=C1)N(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1

Specifications

Quantity 200 mg
Formula Weight 748.89
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-(2,3-Dihydrothieno[3,4-b][1,4]dioxine-5,7-diyl)bis[N,N-bis(4-methoxyphenyl)aniline]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.