Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[[4-(2-butynyloxy)phenyl]sulfonyl]-N-hydroxy-2,2-dimethyl-(3S)thiomorpholinecarboxamide | 287403-39-8 | 99.6% | 398.50 g/mol | C17H22N2O5S2 | 100 MG
SDP

Catalog No. 5000248736
Encompass_Preferred

TMI-1 (WAY-171318) is a small-molecule inhibitor of TNF-α converting enzyme (TACE) with reported activity against ADAM-17, ADAMTS-4, and multiple MMPs. Supplied for research use, it has demonstrated oral activity and induces apoptosis in cancer cell models, and is provided with analytical documentation for laboratory studies.

  • Purity 99.57%.
  • Molecular formula C17H22N2O5S2.
  • Molecular weight 398.50 g/mol.
  • Available as solid (5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and solution (10 mM in DMSO).
  • Includes datasheet, COA, and SDS documentation.
  • Suitable for biochemical, cell-based, and in vivo oral dosing research (research use only).

Catalog No. 50-002-48736 Supplier Medchemexpress LLC Supplier No. HY101448100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TMI-1 (WAY-171318) is a small-molecule inhibitor of TNF-α converting enzyme (TACE) with reported activity against ADAM-17, ADAMTS-4, and multiple MMPs. Supplied for research use, it has demonstrated oral activity and induces apoptosis in cancer cell models, and is provided with analytical documentation for laboratory studies.

  • Purity 99.57%.
  • Molecular formula C17H22N2O5S2.
  • Molecular weight 398.50 g/mol.
  • Available as solid (5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and solution (10 mM in DMSO).
  • Includes datasheet, COA, and SDS documentation.
  • Suitable for biochemical, cell-based, and in vivo oral dosing research (research use only).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.