Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4',4,4'-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid)

Catalog No. 379077250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-907-7250MG 250 mg
37-907-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-907-7250MG Supplier Sigma Aldrich Supplier No. 379077250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4′,4, 4-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid) (H2TCPP) can be used to fabricate three dimensional Ru polymer complex.[1] It can be grafted on SiO2/Nb2O5 substrate and subsequently metallized to investigate catalytic oxidation of hydrazine.

Specifications

CAS 14609-54-2
Molecular Formula C48H30N4O8
Linear Formula C48H30N4O8
MDL Number MFCD00064860
Quantity 250 mg
Synonym meso-Tetraphenylporphine-4,4 ,4,4-tetracarboxylic acid
Molecular Weight (g/mol) 790.77
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.