Learn More
Sigma Aldrich 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzamide
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Methyl 4,4-dimethyl-3-oxopentanoate has been used:
- As substarte during incl3-catalyzed syntheses of some new dibenzo(d,f)(1,3)dioxepines and 12h-dibenzo(d,g)(1,3)dioxocin derivatives
- In the preparation of neat α-oxoketene, carbomethoxypivaloylketene
Specifications
Specifications
| CAS | 179117-44-3 |
| Molecular Formula | C13H18BNO3 |
| Linear Formula | C13H18BNO3 |
| MDL Number | MFCD05663887 |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 247.1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.