Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl Acetate 96.0+%, TCI America™
SDP

Catalog No. T32241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32241G 1 g
T32245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32241G Supplier TCI America Supplier No. T32241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 480424-70-2
Molecular Formula C14H19BO4
Molecular Weight (g/mol) 262.11
MDL Number MFCD03453060
InChI Key KHBAJCWEQNVCSN-UHFFFAOYSA-N
Synonym 4-Acetoxyphenylboronic Acid Pinacol Ester, 2-(4-Acetoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2774004
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate
SMILES CC(=O)OC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 66°C
Color White
Quantity 1 g
Formula Weight 262.11
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.