Learn More
Medchemexpress LLC 4-[[4-(4-chlorophenoxy)phenyl]sulfonylmethyl]-N-hydroxyoxane-4-carboxamide | 193022-04-7 | MFCD16657131 | 99.2% | 425.88 | C19H20ClNO6S | 1 ML
SDP
Catalog No. 5000243313
Encompass_Preferred
CTS-1027 is a potent small-molecule inhibitor of matrix metalloproteinases (MMPs), exhibiting sub-nanomolar potency against MMP2 and MMP13 and strong selectivity over MMP1. The compound is provided with high purity and is available as a 10 mM solution in DMSO (1 mL) or as solid vials in multiple sizes for biochemical and cellular research.
  • Sub-nanomolar potency against MMP2 and MMP13 (IC50 ≈ 0.3 nM and 0.5 nM)
  • Greater than 1,000-fold selectivity over MMP1
  • High purity suitable for research applications
  • Available as 10 mM solution in DMSO for immediate use
  • Supplied in multiple solid vial sizes for flexible dosing and storage
  • Molecular weight 425.88 and formula C19H20ClNO6S for compound identification
Catalog No. 50-002-43313 Supplier Medchemexpress LLC Supplier No. HY1039810MM 1ML
CTS-1027 is a potent small-molecule inhibitor of matrix metalloproteinases (MMPs), exhibiting sub-nanomolar potency against MMP2 and MMP13 and strong selectivity over MMP1. The compound is provided with high purity and is available as a 10 mM solution in DMSO (1 mL) or as solid vials in multiple sizes for biochemical and cellular research.
  • Sub-nanomolar potency against MMP2 and MMP13 (IC50 ≈ 0.3 nM and 0.5 nM)
  • Greater than 1,000-fold selectivity over MMP1
  • High purity suitable for research applications
  • Available as 10 mM solution in DMSO for immediate use
  • Supplied in multiple solid vial sizes for flexible dosing and storage
  • Molecular weight 425.88 and formula C19H20ClNO6S for compound identification