Learn More
Medchemexpress LLC 4-[[4-(4-chlorophenoxy)phenyl]sulfonylmethyl]-N-hydroxyoxane-4-carboxamide | 193022-04-7 | MFCD16657131 | 99.2% | 425.88 | C19H20ClNO6S | 25 MG
SDP
Catalog No. 5000243576
Encompass_Preferred
CTS-1027 is a potent small-molecule inhibitor of matrix metalloproteinases (MMPs) with sub-nanomolar activity against MMP2 and MMP13 and strong selectivity over MMP1. It has been characterized in preclinical models for effects on hepatocyte apoptosis and fibrogenesis and is supplied for research use. The compound is a crystalline solid with high DMSO solubility and defined storage recommendations for powder and solutions.
  • Potent inhibition of MMP2 and MMP13 at sub-nanomolar concentrations.
  • Broad MMP activity including MMP12, MMP8, MMP3, and MMP14.
  • Greater than 1,000-fold selectivity over MMP1.
  • High solubility in DMSO enables concentrated stock preparation.
  • Stable as powder under recommended cold storage conditions.
  • Demonstrated activity in preclinical models of liver injury.
Catalog No. 50-002-43576 Supplier Medchemexpress LLC Supplier No. HY1039825MG
CTS-1027 is a potent small-molecule inhibitor of matrix metalloproteinases (MMPs) with sub-nanomolar activity against MMP2 and MMP13 and strong selectivity over MMP1. It has been characterized in preclinical models for effects on hepatocyte apoptosis and fibrogenesis and is supplied for research use. The compound is a crystalline solid with high DMSO solubility and defined storage recommendations for powder and solutions.
  • Potent inhibition of MMP2 and MMP13 at sub-nanomolar concentrations.
  • Broad MMP activity including MMP12, MMP8, MMP3, and MMP14.
  • Greater than 1,000-fold selectivity over MMP1.
  • High solubility in DMSO enables concentrated stock preparation.
  • Stable as powder under recommended cold storage conditions.
  • Demonstrated activity in preclinical models of liver injury.