Learn More
Medchemexpress LLC 4-[[4-(4-chlorophenoxy)phenyl]sulfonylmethyl]-N-hydroxyoxane-4-carboxamide | 193022-04-7 | MFCD16657131 | 99.2% | 425.88 g/mol | C19H20ClNO6S | 10 MG
SDP
Catalog No. 5000244144
Encompass_Preferred
CTS-1027 is a potent small-molecule inhibitor of matrix metalloproteinases (MMPs) with sub-nanomolar activity against MMP2 and MMP13 and strong selectivity over MMP1. It is supplied as a high-purity research reagent for biochemical and cell-based studies investigating MMP-related pathways.
  • Inhibits MMP2 with IC50 0.3 nM.
  • Inhibits MMP13 with IC50 0.5 nM.
  • Shows >1,000-fold selectivity over MMP1.
  • High purity (99.24%).
  • Powder stable at -20°C for long-term storage.
  • Available in small milligram quantities and as a DMSO solution.
Catalog No. 50-002-44144 Supplier Medchemexpress LLC Supplier No. HY1039810MG
CTS-1027 is a potent small-molecule inhibitor of matrix metalloproteinases (MMPs) with sub-nanomolar activity against MMP2 and MMP13 and strong selectivity over MMP1. It is supplied as a high-purity research reagent for biochemical and cell-based studies investigating MMP-related pathways.
  • Inhibits MMP2 with IC50 0.3 nM.
  • Inhibits MMP13 with IC50 0.5 nM.
  • Shows >1,000-fold selectivity over MMP1.
  • High purity (99.24%).
  • Powder stable at -20°C for long-term storage.
  • Available in small milligram quantities and as a DMSO solution.