Learn More
Medchemexpress LLC 4-[[4-(4-chlorophenoxy)phenyl]sulfonylmethyl]-N-hydroxyoxane-4-carboxamide | 193022-04-7 | MFCD16657131 | 99.2% | 425.88 g/mol | C19H20ClNO6S | 100 MG
SDP
Catalog No. 5000243879
Encompass_Preferred
CTS-1027 is a potent small-molecule matrix metalloproteinase (MMP) inhibitor used for biochemical and cellular research. It exhibits sub-nanomolar activity against key MMP isoforms and is employed to probe MMP-mediated biology and validate therapeutic hypotheses in vitro and in vivo.
  • Sub-nanomolar potency against MMP2 (IC50 0.3 nM) and MMP13 (IC50 0.5 nM).
  • Greater than 1,000-fold selectivity over MMP1.
  • High purity suitable for research (LCMS purity 99.24%).
  • Molecular weight 425.88 g/mol and formula C19H20ClNO6S.
  • Solid, white to off-white appearance for straightforward handling.
Catalog No. 50-002-43879 Supplier Medchemexpress LLC Supplier No. HY10398100MG
CTS-1027 is a potent small-molecule matrix metalloproteinase (MMP) inhibitor used for biochemical and cellular research. It exhibits sub-nanomolar activity against key MMP isoforms and is employed to probe MMP-mediated biology and validate therapeutic hypotheses in vitro and in vivo.
  • Sub-nanomolar potency against MMP2 (IC50 0.3 nM) and MMP13 (IC50 0.5 nM).
  • Greater than 1,000-fold selectivity over MMP1.
  • High purity suitable for research (LCMS purity 99.24%).
  • Molecular weight 425.88 g/mol and formula C19H20ClNO6S.
  • Solid, white to off-white appearance for straightforward handling.