Learn More
Medchemexpress LLC 4-[[4-(4-chlorophenoxy)phenyl]sulfonylmethyl]-N-hydroxyoxane-4-carboxamide | 193022-04-7 | MFCD16657131 | 99.2% | 425.88 g/mol | C19H20ClNO6S | 50 MG
SDP
Catalog No. 5000243314
Encompass_Preferred
CTS-1027 (CAS 193022-04-7) is a small-molecule inhibitor of matrix metalloproteinases for research applications. It potently inhibits MMP2 and MMP13 at sub-nanomolar IC50s and exhibits strong selectivity over MMP1. The compound is supplied as a solid with documented analytical data to support experimental use.
  • Potent inhibition of MMP2 and MMP13 at sub-nanomolar concentrations.
  • High selectivity over MMP1 (>1,000-fold) to reduce off-target activity.
  • High reported chemical purity for reliable experimental results.
  • Provided as a solid suitable for storage and solution preparation.
  • Documented analytical data (COA, LC-MS) available to support characterization.
Catalog No. 50-002-43314 Supplier Medchemexpress LLC Supplier No. HY1039850MG
CTS-1027 (CAS 193022-04-7) is a small-molecule inhibitor of matrix metalloproteinases for research applications. It potently inhibits MMP2 and MMP13 at sub-nanomolar IC50s and exhibits strong selectivity over MMP1. The compound is supplied as a solid with documented analytical data to support experimental use.
  • Potent inhibition of MMP2 and MMP13 at sub-nanomolar concentrations.
  • High selectivity over MMP1 (>1,000-fold) to reduce off-target activity.
  • High reported chemical purity for reliable experimental results.
  • Provided as a solid suitable for storage and solution preparation.
  • Documented analytical data (COA, LC-MS) available to support characterization.