Learn More
Medchemexpress LLC 4-[4,4-dimethyl-8-nitro-7-(4-phenylmethoxycarbonylpiperazin-1-yl)chromeno[4,3-c]pyrazol-1-yl]benzoic | 2024548-03-4 | MFCD30741839 | 99.5% | 583.59 g·mol⁻1 | C31H29N5O7 | 5 MG
SDP
Catalog No. 5000263661
Encompass_Preferred
Lin28-let-7a antagonist 1 is a small-molecule inhibitor that disrupts the interaction between Lin28 proteins and the let-7a microRNA family. The compound demonstrates antagonistic activity against Lin28A-let-7a-1 with a reported IC50 of 4.03 μM and is supplied as a solid or as a 10 mM solution in DMSO for laboratory research.
  • Antagonizes Lin28-let-7a interaction (IC50 4.03 μM).
  • Available as solid packs (5 mg-100 mg) and as 10 mM solution in DMSO.
  • High purity (≈99.5%).
  • Molecular weight 583.59 g·mol⁻1 and formula C31H29N5O7.
  • Light yellow to yellow solid appearance.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-63661 Supplier Medchemexpress LLC Supplier No. HY1006925MG
Lin28-let-7a antagonist 1 is a small-molecule inhibitor that disrupts the interaction between Lin28 proteins and the let-7a microRNA family. The compound demonstrates antagonistic activity against Lin28A-let-7a-1 with a reported IC50 of 4.03 μM and is supplied as a solid or as a 10 mM solution in DMSO for laboratory research.
  • Antagonizes Lin28-let-7a interaction (IC50 4.03 μM).
  • Available as solid packs (5 mg-100 mg) and as 10 mM solution in DMSO.
  • High purity (≈99.5%).
  • Molecular weight 583.59 g·mol⁻1 and formula C31H29N5O7.
  • Light yellow to yellow solid appearance.
  • Intended for research use only; not for human or veterinary use.