Learn More
Medchemexpress LLC 4-(4-(4-fluorophenyl)-2-(4-(methylsulfonyl)phenyl)-1H-imidazol-5-yl)pyridine | 152121-46-5 | 99.3% | 393.43 g·mol⁻¹ | C21H16FN3O2S | 5 MG
SDP
Catalog No. 5000264736
Encompass_Preferred
SB 203580 sulfone is a sulfone analog of the p38 MAP kinase inhibitor used as a research reagent to probe p38 MAPK-mediated signaling and inflammatory responses. It inhibits IL-1 production in monocytes and shows activity in pathways related to 5-lipoxygenase. Available as solid material or as a DMSO solution for assay use.
Catalog No. 50-002-64736 Supplier Medchemexpress LLC Supplier No. HY1123495MG
SB 203580 sulfone is a sulfone analog of the p38 MAP kinase inhibitor used as a research reagent to probe p38 MAPK-mediated signaling and inflammatory responses. It inhibits IL-1 production in monocytes and shows activity in pathways related to 5-lipoxygenase. Available as solid material or as a DMSO solution for assay use.