Learn More
Medchemexpress LLC 4-[4-(4-fluorophenyl)-2-(4-nitrophenyl)-1H-imidazol-5-yl]pyridine | 152121-53-4 | MFCD12405019 | 98.0% | 360.34 | C20H13FN4O2 | 10 MG
SDP
Catalog No. 5000240236
Encompass_Preferred
PD 169316 (Standard) is an analytical reference material intended for research and analytical applications. It corresponds to a selective p38 MAP kinase inhibitor (CAS 152121-53-4) with molecular formula C20H13FN4O2 and molecular weight 360.34. The standard is supplied as a 10 mg solid with reported purity of about 98% and is intended for assay calibration, method validation, and reference testing.
  • Intended for assay calibration and method validation.
  • High reported purity suitable for analytical use.
  • Supplied as a 10 mg solid for convenient handling.
  • Includes molecular formula and molecular weight for reference.
  • Useful for research and analytical workflows involving kinase assays.
Catalog No. 50-002-40236 Supplier Medchemexpress LLC Supplier No. HY10578R10MG
PD 169316 (Standard) is an analytical reference material intended for research and analytical applications. It corresponds to a selective p38 MAP kinase inhibitor (CAS 152121-53-4) with molecular formula C20H13FN4O2 and molecular weight 360.34. The standard is supplied as a 10 mg solid with reported purity of about 98% and is intended for assay calibration, method validation, and reference testing.
  • Intended for assay calibration and method validation.
  • High reported purity suitable for analytical use.
  • Supplied as a 10 mg solid for convenient handling.
  • Includes molecular formula and molecular weight for reference.
  • Useful for research and analytical workflows involving kinase assays.