Learn More
Medchemexpress LLC 4-[4-(4-fluorophenyl)-2-(4-nitrophenyl)-1H-imidazol-5-yl]pyridine | 152121-53-4 | MFCD12405019 | 98.0% | 360.34 g·mol⁻¹ | C20H13FN4O2 | 50 MG
SDP
Catalog No. 5000241554
Encompass_Preferred
PD 169316 is an analytical standard and research reagent used to inhibit p38 mitogen-activated protein kinase in biochemical and cell-based studies. It is a cell-permeable pyridinyl imidazole compound supplied as a high-purity solid for assay development, compound identification, and analytical characterization.
  • High purity (≥98% by HPLC) suitable for analytical use.
  • Selective p38 MAP kinase inhibitor with reported IC50 ≈ 89 nM.
  • Cell-permeable small molecule compatible with biochemical and cellular assays.
  • Molecular weight 360.34 g·mol⁻¹ and defined molecular formula for analytical reference.
  • Available in small pack sizes for analytical and research applications.
Catalog No. 50-002-41554 Supplier Medchemexpress LLC Supplier No. HY10578R50MG
PD 169316 is an analytical standard and research reagent used to inhibit p38 mitogen-activated protein kinase in biochemical and cell-based studies. It is a cell-permeable pyridinyl imidazole compound supplied as a high-purity solid for assay development, compound identification, and analytical characterization.
  • High purity (≥98% by HPLC) suitable for analytical use.
  • Selective p38 MAP kinase inhibitor with reported IC50 ≈ 89 nM.
  • Cell-permeable small molecule compatible with biochemical and cellular assays.
  • Molecular weight 360.34 g·mol⁻¹ and defined molecular formula for analytical reference.
  • Available in small pack sizes for analytical and research applications.