Learn More
Medchemexpress LLC 4-[4-(4-fluorophenyl)-2-(4-nitrophenyl)-1H-imidazol-5-yl]pyridine | 152121-53-4 | MFCD12405019 | 98.6% | 360.34 | C20H13FN4O2 | 500 MG
SDP
Catalog No. 5000240140
Encompass_Preferred
PD 169316 is a potent, cell-permeable, and selective p38 MAP kinase inhibitor used as a research reagent and analytical standard for biochemical and cell-based studies. It inhibits p38 MAPK with a reported IC50 of approximately 89 nM and is supplied in multiple pack sizes for laboratory use.
  • Potent, selective p38 MAP kinase inhibitor.
  • Cell-permeable small molecule suitable for cell-based assays.
  • Reported IC50 approximately 89 nM against p38 MAPK.
  • High purity, typically 98.6% by HPLC.
  • Molecular weight 360.34 g/mol and formula C20H13FN4O2.
  • Provided as an analytical standard in multiple pack sizes, including 500 mg.
  • Intended for research use only, not for human consumption.
Catalog No. 50-002-40140 Supplier Medchemexpress LLC Supplier No. HY10578500MG
PD 169316 is a potent, cell-permeable, and selective p38 MAP kinase inhibitor used as a research reagent and analytical standard for biochemical and cell-based studies. It inhibits p38 MAPK with a reported IC50 of approximately 89 nM and is supplied in multiple pack sizes for laboratory use.
  • Potent, selective p38 MAP kinase inhibitor.
  • Cell-permeable small molecule suitable for cell-based assays.
  • Reported IC50 approximately 89 nM against p38 MAPK.
  • High purity, typically 98.6% by HPLC.
  • Molecular weight 360.34 g/mol and formula C20H13FN4O2.
  • Provided as an analytical standard in multiple pack sizes, including 500 mg.
  • Intended for research use only, not for human consumption.