Learn More
Medchemexpress LLC 4-[4-(4-fluorophenyl)-2-(4-nitrophenyl)-1H-imidazol-5-yl]pyridine | 152121-53-4 | MFCD12405019 | 98.6% | 360.34 g·mol⁻¹ | C20H13FN4O2 | 100 MG
SDP
Catalog No. 5000240835
Encompass_Preferred
PD 169316 is a selective, cell-permeable inhibitor of p38 mitogen-activated protein kinase used in biochemical and cellular research. It exhibits an IC50 of approximately 89 nM, is supplied as a high-purity solid powder, and is stored under low-temperature conditions to preserve stability.
  • Selective p38 MAPK inhibition (IC50 approximately 89 nM).
  • High purity suitable for research applications (reported 98.6%).
  • Supplied as a solid powder for ease of handling and formulation.
  • Molecular weight 360.34 g·mol⁻¹; formula C20H13FN4O2.
  • Stable when stored at recommended low temperatures.
Catalog No. 50-002-40835 Supplier Medchemexpress LLC Supplier No. HY10578100MG
PD 169316 is a selective, cell-permeable inhibitor of p38 mitogen-activated protein kinase used in biochemical and cellular research. It exhibits an IC50 of approximately 89 nM, is supplied as a high-purity solid powder, and is stored under low-temperature conditions to preserve stability.
  • Selective p38 MAPK inhibition (IC50 approximately 89 nM).
  • High purity suitable for research applications (reported 98.6%).
  • Supplied as a solid powder for ease of handling and formulation.
  • Molecular weight 360.34 g·mol⁻¹; formula C20H13FN4O2.
  • Stable when stored at recommended low temperatures.