Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[4-(4-isoquinolinyl)phenyl]-N,N-dimethyl-1H-pyrazole-1-acetamide | 2163056-91-3 | 99.8% | 100 MG
SDP

Catalog No. 5000234200
Encompass_Preferred

BI-1347 is an orally active, selective small-molecule inhibitor of cyclin-dependent kinase 8 (CDK8) used as a preclinical research tool to study CDK8-mediated signaling and antitumor activity. It is supplied as a light yellow solid with high chemical purity and is available in small solid quantities and DMSO solutions for in vitro use.

  • Orally active, selective CDK8 inhibitor with high potency (IC50 ≈ 1.1 nM).
  • Used as a research reagent to study CDK8-mediated signaling and antitumor activity.
  • Light yellow solid with high purity (>99% by HPLC).
  • Molecular formula C22H20N4O; molecular weight 356.42 g·mol⁻1.
  • Available as small solid quantities and 10 mM solutions in DMSO for assay use.
  • Recommended storage: powder at -20°C; solutions at -80°C for long-term storage.

Catalog No. 50-002-34200 Supplier Medchemexpress LLC Supplier No. HY120350100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-1347 is an orally active, selective small-molecule inhibitor of cyclin-dependent kinase 8 (CDK8) used as a preclinical research tool to study CDK8-mediated signaling and antitumor activity. It is supplied as a light yellow solid with high chemical purity and is available in small solid quantities and DMSO solutions for in vitro use.

  • Orally active, selective CDK8 inhibitor with high potency (IC50 ≈ 1.1 nM).
  • Used as a research reagent to study CDK8-mediated signaling and antitumor activity.
  • Light yellow solid with high purity (>99% by HPLC).
  • Molecular formula C22H20N4O; molecular weight 356.42 g·mol⁻1.
  • Available as small solid quantities and 10 mM solutions in DMSO for assay use.
  • Recommended storage: powder at -20°C; solutions at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.