Learn More
Medchemexpress LLC 4-[4-(4-isoquinolinyl)phenyl]-N,N-dimethyl-1H-pyrazole-1-acetamide | 2163056-91-3 | 99.8% | 50 MG
SDP
Catalog No. 5000233510
Encompass_Preferred
BI-1347 is an orally active, selective, and potent inhibitor of cyclin-dependent kinase 8 (CDK8) with sub-nanomolar biochemical potency and demonstrated antitumor activity in preclinical models. It is supplied for research use in solid and solution formats in multiple pack sizes.
  • Orally active small-molecule CDK8 inhibitor.
  • Biochemical IC50 ~1-1.1 nM for CDK8/cyclinC.
  • Molecular formula C22H20N4O; molecular weight 356.42 Da.
  • Supplied purity approximately 99.8%.
  • Available in solid and solution formats and multiple pack sizes including 50 MG.
Catalog No. 50-002-33510 Supplier Medchemexpress LLC Supplier No. HY12035050MG
BI-1347 is an orally active, selective, and potent inhibitor of cyclin-dependent kinase 8 (CDK8) with sub-nanomolar biochemical potency and demonstrated antitumor activity in preclinical models. It is supplied for research use in solid and solution formats in multiple pack sizes.
  • Orally active small-molecule CDK8 inhibitor.
  • Biochemical IC50 ~1-1.1 nM for CDK8/cyclinC.
  • Molecular formula C22H20N4O; molecular weight 356.42 Da.
  • Supplied purity approximately 99.8%.
  • Available in solid and solution formats and multiple pack sizes including 50 MG.