Learn More
Medchemexpress LLC 4,4'-(4-methyl-5-(4-(2-(piperidin-1-yl)ethoxy)phenyl)-1H-pyrazole-1,3-diyl)diphenol hydrochloride | 2863676-89-3 | MFCD34676763 | 99.3% | 506.04 g/mol | C29H32ClN3O3 | 1 ML
SDP
Catalog No. 5000247253
Encompass_Preferred
MPP hydrochloride is a potent, selective estrogen receptor (ER) modulator used in in vitro and in vivo research to probe ERα activity. It is supplied as a white to off-white solid and as a ready-to-use 10 mM solution in DMSO (1 mL). Reported purity is 99.28% and molecular weight is 506.04 g/mol (C29H32ClN3O3).
  • Potent, selective ER modulator for ERα studies.
  • Available as a solid or 10 mM solution in DMSO.
  • High reported purity (99.28%).
  • Suitable for cellular and animal model experiments.
  • Storage recommendations for solid and solution provided.
Catalog No. 50-002-47253 Supplier Medchemexpress LLC Supplier No. HY103454B10MM 1ML
MPP hydrochloride is a potent, selective estrogen receptor (ER) modulator used in in vitro and in vivo research to probe ERα activity. It is supplied as a white to off-white solid and as a ready-to-use 10 mM solution in DMSO (1 mL). Reported purity is 99.28% and molecular weight is 506.04 g/mol (C29H32ClN3O3).
  • Potent, selective ER modulator for ERα studies.
  • Available as a solid or 10 mM solution in DMSO.
  • High reported purity (99.28%).
  • Suitable for cellular and animal model experiments.
  • Storage recommendations for solid and solution provided.