Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4',4″-Tris(4,5-dichlorophthalimido)trityl Bromide [Protecting Reagent for Primary Alcohol], TCI America™
SDP

Catalog No. T15261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15261G Supplier TCI America Supplier No. T15261G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 91898-93-0
Molecular Formula C43H18BrCl6N3O6
Molecular Weight (g/mol) 965.24
MDL Number MFCD00075041
InChI Key JYHNGHXQORJDGQ-UHFFFAOYSA-N
Synonym p,p′C,p′C′C-(Bromomethylidyne)tris(4,5-dichloro-N-phenylphthalimide)
PubChem CID 3623731
IUPAC Name 2-(4-{bromobis[4-(5,6-dichloro-1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)phenyl]methyl}phenyl)-5,6-dichloro-2,3-dihydro-1H-isoindole-1,3-dione
SMILES ClC1=C(Cl)C=C2C(=O)N(C(=O)C2=C1)C1=CC=C(C=C1)C(Br)(C1=CC=C(C=C1)N1C(=O)C2=CC(Cl)=C(Cl)C=C2C1=O)C1=CC=C(C=C1)N1C(=O)C2=CC(Cl)=C(Cl)C=C2C1=O

Specifications

Quantity 1 g
UN Number 1759
Formula Weight 965.24
Chemical Name or Material 4,4′,4′′-Tris(4,5-dichlorophthalimido)trityl Bromide [Protecting Reagent for Prima
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.