Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4',4″-Tris[phenyl(m-tolyl)amino]triphenylamine 98.0+%, TCI America™
SDP

Catalog No. T2251100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2251100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2251100MG Supplier TCI America Supplier No. T2251100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 124729-98-2
Molecular Formula C57H48N4
Molecular Weight (g/mol) 789.039
InChI Key DIVZFUBWFAOMCW-UHFFFAOYSA-N
Synonym m-MTDATA
PubChem CID 11061735
IUPAC Name 4-N-(3-methylphenyl)-1-N,1-N-bis[4-(N-(3-methylphenyl)anilino)phenyl]-4-N-phenylbenzene-1,4-diamine
SMILES CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)N(C4=CC=C(C=C4)N(C5=CC=CC=C5)C6=CC=CC(=C6)C)C7=CC=C(C=C7)N(C8=CC=CC=C8)C9=CC=CC(=C9)C

Specifications

Melting Point 210°C
Color Yellow
Quantity 100 mg
Formula Weight 789.04
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Tris[phenyl(m-tolyl)amino]triphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.