Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,5,5,5-Pentafluoropentanoic acid, Thermo Scientific™

Catalog No. MAY00177DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAY00177DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAY00177DA Supplier Thermo Scientific Chemicals Supplier No. MAY00177DA
Add to Cart

Chemical Identifiers

CAS 3637-31-8
Molecular Formula C5H5F5O2
Molecular Weight (g/mol) 192.085
InChI Key HHSIXPRDVJARSI-UHFFFAOYSA-N
Synonym pentanoic acid, 4,4,5,5,5-pentafluoro, pentanoicacid,4,4,5,5,5-pentafluoro
PubChem CID 23423163
IUPAC Name 4,4,5,5,5-pentafluoropentanoic acid
SMILES C(CC(C(F)(F)F)(F)F)C(=O)O

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 192.09
Percent Purity 90%
Chemical Name or Material 4,4,5,5,5-Pentafluoropentanoic acid
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.