Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,5,5,5-Pentafluoropentanol, 94%

Catalog No. p-7025823
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2151204 2 g
AAB2151209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2151204 Supplier Thermo Scientific Chemicals Supplier No. B2151204
Only null left
Add to Cart

CAS: 148043-73-6 | C5H7F5O | 178.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148043-73-6
Molecular Formula C5H7F5O
Molecular Weight (g/mol) 178.102
MDL Number MFCD00153224
InChI Key QROUUECTKRZFHF-UHFFFAOYSA-N
Synonym 4,4,5,5,5-pentafluoro-1-pentanol, 4,4,5,5,5-pentafluoropentanol, pentafluoropentanol, 1-pentanol, 4,4,5,5,5-pentafluoro, fulvestrant intermediate, acmc-1bz8x, 3-perfluoroethyl propan-1-ol, 4,4,5,5,5-pentafluoro pentanol, 4,4,5,5,5-pentafluoro-pentanol, 4,4,5,5,5-pentafluorpentan-1-ol
PubChem CID 547967
IUPAC Name 4,4,5,5,5-pentafluoropentan-1-ol
SMILES C(CC(C(F)(F)F)(F)F)CO

Specifications

Density 1.35
Boiling Point 135°C
Flash Point 63°C (145°F)
Refractive Index 1.334
Linear Formula CF3CF2(CH2)3OH
Quantity 2 g
Formula Weight 178.1
Percent Purity 94%
Chemical Name or Material 4,4,5,5,5-Pentafluoropentanol
Hazard Category H227-H302
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 421-360-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.