Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane 98.0+%, TCI America™
SDP

Catalog No. T34695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34695G 5 g
T34691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34695G Supplier TCI America Supplier No. T34695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 126689-00-7
Molecular Formula C10H19BO2
Molecular Weight (g/mol) 182.07
MDL Number MFCD09842762
InChI Key LWXHOCHDERDUID-UHFFFAOYSA-N
Synonym (2-Methyl-1-propenyl)boronic Acid Pinacol Ester
PubChem CID 15148447
IUPAC Name 4,4,5,5-tetramethyl-2-(2-methylprop-1-enyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=C(C)C

Specifications

Color Yellow
Boiling Point 71°C
Quantity 5 g
Formula Weight 182.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(2-methyl-1-propenyl)-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.