Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,5,5-Tetramethyl-2-(m-tolyl)-1,3,2-dioxaborolane 98.0+%, TCI America™
SDP

Catalog No. T32195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32195G 5 g
T32191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32195G Supplier TCI America Supplier No. T32195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 253342-48-2
Molecular Formula C13H19BO2
Molecular Weight (g/mol) 218.10
MDL Number MFCD05864303
InChI Key XDKYCZBGHPGKEP-UHFFFAOYSA-N
Synonym 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene, m-Tolylboronic Acid Pinacol Ester
PubChem CID 2760598
IUPAC Name 4,4,5,5-tetramethyl-2-(3-methylphenyl)-1,3,2-dioxaborolane
SMILES CC1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 36°C
Color White
Quantity 5 g
Formula Weight 218.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(m-tolyl)-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.