Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4,5,5,-Tetramethyl-2-Phenylsulfanylmethyl-1,3,2-Dioxaborolane

Catalog No. 5681555G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-815-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-815-55G Supplier Sigma Aldrich Supplier No. 5681555G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4,5,5,-Tetramethyl-2-phenylsulfanylmethyl-1,3,2-dioxaborolane is used to prepare aryl/heteroaryl derivatives via the formation of C-C and C-hetero bonds.It can also be used as:A starting material in the synthesis of +a-aminoboronic acids as serine proteases inhibitors.A substrate in the coupling reactions with carbonyl compounds under Ir-catalyzed photoredox conditions.

Specifications

CAS 4334-88-7
Melting Point 135°C
Molecular Formula C9H11BO4
Refractive Index n20/D 1.5280
Linear Formula C2H5OCOC6H4B(OH)2
MDL Number MFCD02179441
Quantity 5 g
Synonym 4-Boronobenzoic acid 1-ethyl ester; 4-Carboethoxybenzeneboronic acid; Ethyl 4-boronobenzoate
Molecular Weight (g/mol) 193.99 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.