Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[(4,5-dihydro-3-isoxazolyl)oxy]-N,N,N-trimethyl-2-butyn-1-aminium iodide | 247079-84-1 | 99.5% | 1 ML
SDP

Catalog No. 5000230407
Encompass_Preferred

Iperoxo is a potent muscarinic acetylcholine receptor (mAChR) superagonist provided for research use, supplied as a 10 mM solution in DMSO and in solid quantities for in vitro pharmacology and receptor activation studies.

  • Potent mAChR superagonist for receptor activation studies.
  • Provided as 10 mM solution in DMSO, 1 mL vial.
  • High purity (≈99.5%) suitable for biochemical assays.
  • Molecular formula C10H17IN2O2; molecular weight 324.16 g/mol.
  • Available as solid quantities for formulation flexibility.

Catalog No. 50-002-30407 Supplier Medchemexpress LLC Supplier No. HY12274310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iperoxo is a potent muscarinic acetylcholine receptor (mAChR) superagonist provided for research use, supplied as a 10 mM solution in DMSO and in solid quantities for in vitro pharmacology and receptor activation studies.

  • Potent mAChR superagonist for receptor activation studies.
  • Provided as 10 mM solution in DMSO, 1 mL vial.
  • High purity (≈99.5%) suitable for biochemical assays.
  • Molecular formula C10H17IN2O2; molecular weight 324.16 g/mol.
  • Available as solid quantities for formulation flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.