Learn More
Medchemexpress LLC 4-(4,5-diphenyl-1H-imidazol-2-yl)benzoic acid | 5496-35-5 | MFCD00999654 | 99.7% | 340.37 | C22H16N2O2 | 250 MG
SDP
Catalog No. 5000246369
Encompass_Preferred
I-XW-053 is a small-molecule inhibitor of the HIV-1 capsid protein used in antiviral research; it disrupts the capsid N-terminal domain (NTD-NTD) interface and exhibits micromolar-range activity.
  • Inhibits HIV-1 capsid protein by blocking the NTD-NTD interface.
  • Shows micromolar antiviral activity (reported IC50 ≈ 164.2 μM).
  • High purity suitable for research applications.
  • Available in multiple pack sizes for flexible experimental needs.
  • Solid form; soluble in DMSO for solution preparation.
Catalog No. 50-002-46369 Supplier Medchemexpress LLC Supplier No. HY103078250MG
I-XW-053 is a small-molecule inhibitor of the HIV-1 capsid protein used in antiviral research; it disrupts the capsid N-terminal domain (NTD-NTD) interface and exhibits micromolar-range activity.
  • Inhibits HIV-1 capsid protein by blocking the NTD-NTD interface.
  • Shows micromolar antiviral activity (reported IC50 ≈ 164.2 μM).
  • High purity suitable for research applications.
  • Available in multiple pack sizes for flexible experimental needs.
  • Solid form; soluble in DMSO for solution preparation.