Learn More
Medchemexpress LLC 4-(4,5-diphenyl-1H-imidazol-2-yl)benzoyl chloride | 162756-62-9 | 98.0% | 358.82 | C22H15ClN2O | 10 MG
SDP
Catalog No. 5000297314
Encompass_Preferred
4-(4,5-diphenyl-1H-imidazol-2-yl)benzoyl chloride is a benzoyl chloride derivative used as a labeling reagent for amines and for HPLC derivatization in analytical and research workflows. Supplied as a solid with high purity suitable for laboratory use.
  • Used to derivatize amines for HPLC analysis
  • Fluorescent imidazolyl moiety aids sensitive detection
  • High purity (98.0%) suitable for analytical work
  • Available in small, controlled package sizes for research use
  • Stable solid form for convenient storage and handling
Catalog No. 50-002-97314 Supplier Medchemexpress LLC Supplier No. HYD005910MG
4-(4,5-diphenyl-1H-imidazol-2-yl)benzoyl chloride is a benzoyl chloride derivative used as a labeling reagent for amines and for HPLC derivatization in analytical and research workflows. Supplied as a solid with high purity suitable for laboratory use.
  • Used to derivatize amines for HPLC analysis
  • Fluorescent imidazolyl moiety aids sensitive detection
  • High purity (98.0%) suitable for analytical work
  • Available in small, controlled package sizes for research use
  • Stable solid form for convenient storage and handling