Learn More
Medchemexpress LLC 4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoyl chloride | 162756-62-9 | 98.0% | 5 MG
SDP
Catalog No. 5000410704
Encompass_Preferred
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoyl chloride (DIB-Cl) is a fluorescent labeling reagent. It can be used for labeling amines and has been successfully applied to determine activator amines by HPLC.
  • Fluorescent labeling reagent
  • Can be used for labeling amines
  • Successfully applied to determine activator amines by HPLC
Catalog No. 50-004-10704 Supplier Medchemexpress LLC Supplier No. HYD00595MG
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoyl chloride (DIB-Cl) is a fluorescent labeling reagent. It can be used for labeling amines and has been successfully applied to determine activator amines by HPLC.
  • Fluorescent labeling reagent
  • Can be used for labeling amines
  • Successfully applied to determine activator amines by HPLC