Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(4-amino-3-fluorophenoxy)-N-methylpicolinamide | 757251-39-1 | MFCD09033852 | >95% | C13H12FN3O2 | 50 MG
SDP

Catalog No. 5000434554
Encompass_Preferred

4-(4-Amino-3-fluorophenoxy)-N-methylpicolinamide is a chemical compound primarily used as an analytical standard for research and analytical applications. This substance is identified by its unique CAS number and molecular formula, making it suitable for various laboratory studies and analyses.

  • Identified as an analytical standard
  • Intended for research and analytical applications
  • CAS number: 757251-39-1
  • Molecular formula: C13H12FN3O2
  • Molecular weight: 261.26

Catalog No. 50-004-34554 Supplier Medchemexpress LLC Supplier No. HY7782950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(4-Amino-3-fluorophenoxy)-N-methylpicolinamide is a chemical compound primarily used as an analytical standard for research and analytical applications. This substance is identified by its unique CAS number and molecular formula, making it suitable for various laboratory studies and analyses.

  • Identified as an analytical standard
  • Intended for research and analytical applications
  • CAS number: 757251-39-1
  • Molecular formula: C13H12FN3O2
  • Molecular weight: 261.26

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.