Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Aminophenoxy)-N-methyl-2-pyridinecarboxamide 98.0+%, TCI America™
SDP

Catalog No. A26175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26175G 5 g
A26171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26175G Supplier TCI America Supplier No. A26175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 284462-37-9
Molecular Formula C13H13N3O2
Molecular Weight (g/mol) 243.27
MDL Number MFCD08692047
InChI Key RXZZBPYPZLAEFC-UHFFFAOYSA-N
Synonym 4-(4-Aminophenoxy)-N-methylpicolinamide
PubChem CID 16655129
IUPAC Name 4-(4-aminophenoxy)-N-methylpyridine-2-carboxamide
SMILES CNC(=O)C1=NC=CC(OC2=CC=C(N)C=C2)=C1

Specifications

Melting Point 118°C
Quantity 5 g
Formula Weight 243.27
Percent Purity ≥98.0% (HPLC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Aminophenoxy)-N-methyl-2-pyridinecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.