Learn More
Medchemexpress LLC 4-(4-Aminophenyl)morpholin-3-one | 438056-69-0 | 100.0% | C10H12N2O2 | 1 G
SDP
Catalog No. 5000357815
Encompass_Preferred
4-(4-Aminophenyl)morpholin-3-one is a solid drug intermediate used for the synthesis of various active compounds. It is intended for research purposes only and not for human use. This product has not been fully validated for medical applications.
  • Solid, white to light brown appearance
  • Stable under recommended storage conditions
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Shipment at room temperature if less than 2 weeks
Catalog No. 50-003-57815 Supplier Medchemexpress LLC Supplier No. HY501871G
4-(4-Aminophenyl)morpholin-3-one is a solid drug intermediate used for the synthesis of various active compounds. It is intended for research purposes only and not for human use. This product has not been fully validated for medical applications.
  • Solid, white to light brown appearance
  • Stable under recommended storage conditions
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Shipment at room temperature if less than 2 weeks