Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(4-Aminophenyl)morpholin-3-one | 438056-69-0 | 100.0% | C10H12N2O2 | 50 G
SDP

Catalog No. 5000358096
Encompass_Preferred

4-(4-Aminophenyl)morpholin-3-one is a drug intermediate for the synthesis of various active compounds. This product is a high-purity chemical useful in various research and development settings.

  • Molecular weight: 192.22
  • Molecular formula: C10H12N2O2
  • CAS number: 438056-69-0
  • Appearance: Solid, white to light brown
  • Purity: 100.0%
  • Ships at room temperature in the continental US
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-58096 Supplier Medchemexpress LLC Supplier No. HY5018750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-(4-Aminophenyl)morpholin-3-one is a drug intermediate for the synthesis of various active compounds. This product is a high-purity chemical useful in various research and development settings.

  • Molecular weight: 192.22
  • Molecular formula: C10H12N2O2
  • CAS number: 438056-69-0
  • Appearance: Solid, white to light brown
  • Purity: 100.0%
  • Ships at room temperature in the continental US
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.