Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(4-Aminophenyl)morpholin-3-one | 438056-69-0 | 99.97% | C10H12N2O2 | 500 G
SDP

Catalog No. 5000359209
Encompass_Preferred

4-(4-Aminophenyl)morpholin-3-one is a drug intermediate for the synthesis of various active compounds.

  • Appearance: Solid
  • Color: White to light brown
  • Recommended powder storage: -20°C for 3 years, 4°C for 2 years
  • Recommended in-solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-59209 Supplier Medchemexpress LLC Supplier No. HY50187500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(4-Aminophenyl)morpholin-3-one is a drug intermediate for the synthesis of various active compounds.

  • Appearance: Solid
  • Color: White to light brown
  • Recommended powder storage: -20°C for 3 years, 4°C for 2 years
  • Recommended in-solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.