Learn More
Medchemexpress LLC 4-(4-Aminophenyl)morpholin-3-one | 438056-69-0 | C10H12N2O2 | 10 G
SDP
Supplier:  Medchemexpress LLC HY5018710G
Encompass_Preferred
4-(4-Aminophenyl)morpholin-3-one is a drug intermediate used for the synthesis of various active compounds. As a powder, it can be stored at -20°C for up to 3 years or at 4°C for up to 2 years. When in solvent, it maintains stability at -80°C for 6 months or -20°C for 1 month.
  • Harmful if swallowed.
  • Causes skin irritation.
  • Causes serious eye irritation.
  • May cause respiratory irritation.
Catalog No. 50-003-58824