Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(4-Aminophenyl)morpholin-3-one | 438056-69-0 | C10H12N2O2 | 25 G
SDP

Catalog No. 5000358517
Encompass_Preferred

4-(4-Aminophenyl)morpholin-3-one is a chemical compound used as a drug intermediate for the synthesis of various active compounds. It is stable under recommended storage conditions, ensuring product integrity for research and development applications.

  • Used as a drug intermediate
  • Suitable for synthesis of active compounds
  • Stable for 3 years at -20°C as powder
  • Stable for 2 years at 4°C as powder
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent
  • Melting point between 174.0-174.4°C

Catalog No. 50-003-58517 Supplier Medchemexpress LLC Supplier No. 25GHY50187
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(4-Aminophenyl)morpholin-3-one is a chemical compound used as a drug intermediate for the synthesis of various active compounds. It is stable under recommended storage conditions, ensuring product integrity for research and development applications.

  • Used as a drug intermediate
  • Suitable for synthesis of active compounds
  • Stable for 3 years at -20°C as powder
  • Stable for 2 years at 4°C as powder
  • Stable for 6 months at -80°C in solvent
  • Stable for 1 month at -20°C in solvent
  • Melting point between 174.0-174.4°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.