Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4'-Azobis(4-cyanovaleric acid)

Catalog No. 1159025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-590-25G 25 g
11-590-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-590-25G Supplier Sigma Aldrich Supplier No. 1159025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4'-Azobis(4-cyanovaleric acid)

Specifications

Melting Point 118°C to 125°C (dec.) (lit.)
CAS 2638-94-0
Molecular Formula C12H16N4O4
Linear Formula HOCOCH2CH2C(CH3)(CN)N=NC(CH3)(CN)CH2CH2COOH
MDL Number MFCD00002799
Synonym 4,4 -Azobis(4-cyanopentanoic acid); ABCVA; ACVA
Quantity 25 g
Molecular Weight (g/mol) 280.28
Percent Purity ≥98.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.