Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Azoxydiphenetole 98.0+%, TCI America™
SDP

Catalog No. A06831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06831G 1 g
A06835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06831G Supplier TCI America Supplier No. A06831G

Chemical Identifiers

CAS 4792-83-0
Molecular Formula C16H18N2O3
Molecular Weight (g/mol) 286.33
MDL Number MFCD00026790
InChI Key QUICZVHSJNKDBL-UHFFFAOYSA-N
PubChem CID 78520
IUPAC Name 1,2-bis(4-ethoxyphenyl)-2-oxohydrazin-2-ium-1-ide
SMILES CCOC1=CC=C([N-][N+](=O)C2=CC=C(OCC)C=C2)C=C1

Specifications

Melting Point 132°C
Quantity 1 g
Synonym 4,4′C-Diethoxyazoxybenzene
Formula Weight 286.33
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Azoxydiphenetole
RTECSNumber HM2966400
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.