Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4,4'-Biphenyldiboronic Acid

Catalog No. 456799500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-679-9500MG 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-679-9500MG Supplier Sigma Aldrich Supplier No. 456799500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4,4'-Biphenyldiboronic Acid

Specifications

CAS 1676-81-9
Melting Point 41°C to 43°C
Boiling Point 170°C (0.01 mmHg, lit.)
Molecular Formula C12H15NO5
Linear Formula HOCH2CH(NHCO2CH2C6H5)CO2CH3
MDL Number MFCD00077039
Quantity 10 g
Molecular Weight (g/mol) 253.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.