Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Biphenyldicarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. B28545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28545G 5 g
B28541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28545G Supplier TCI America Supplier No. B28545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66-98-8
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
InChI Key FEHLIYXNTWAEBQ-UHFFFAOYSA-N
Synonym 4,4'-biphenyldicarboxaldehyde, 1,1'-biphenyl-4,4'-dicarbaldehyde, 4,4'-diformylbiphenyl, biphenyl-4,4'-dicarbaldehyde, 4,4'-biphenyldicarboxyaldehyde, 1,1'-biphenyl-4,4'-dicarboxaldehyde, p,p'-diformylbiphenyl, 4-4-formylphenyl benzaldehyde, 4,4'-bis formyl biphenyl
PubChem CID 6200
IUPAC Name 4-(4-formylphenyl)benzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=CC=C(C=C2)C=O

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 5 g
Formula Weight 210.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldicarboxaldehyde
RTECSNumber DV2800200
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.