Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Biphthalic Anhydride (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. B42625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B42625G Supplier TCI America Supplier No. B42625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity monomer used as a raw material for polyimide synthesis.

Chemical Identifiers

CAS 2420-87-3
Molecular Formula C16H6O6
Molecular Weight (g/mol) 294.218
MDL Number MFCD00039140
InChI Key WKDNYTOXBCRNPV-UHFFFAOYSA-N
Synonym Diphenyl-3,3′C,4,4′C-tetracarboxylic Dianhydride
PubChem CID 75494
IUPAC Name 5-(1,3-dioxo-2-benzofuran-5-yl)-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 294.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphthalic Anhydride (purified by sublimation)
EINECSNumber (4)-0833
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.