Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bipyridyl 98.0+%, TCI America™
SDP

Catalog No. B0469100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0469100G 100 g
B046925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0469100G Supplier TCI America Supplier No. B0469100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 553-26-4
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.19
MDL Number MFCD00006416
InChI Key MWVTWFVJZLCBMC-UHFFFAOYSA-N
Synonym 4,4'-bipyridine, 4,4'-bipyridyl, 4,4'-dipyridyl, 4-4-pyridyl pyridine, 4,4'-dipyridine, 4,4-bipyridyl, 4,4-dipyridyl, gamma,gamma'-bipyridyl, gamma,gamma'-dipyridyl, unii-x4o2od61cb
PubChem CID 11107
ChEBI CHEBI:30985
IUPAC Name 4,4'-bipyridine
SMILES C1=CC(=CC=N1)C1=CC=NC=C1

Specifications

Melting Point 112°C
Color White-Yellow
Boiling Point 161°C
Quantity 100 g
UN Number 2811
Formula Weight 156.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bipyridyl
EINECSNumber (5)-3723
RTECSNumber DW1760000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.