Learn More
Strem, An Ascensus Company CAS# 2042201-18-1. 250mg. [4,4'-Bis(1,1-dimethylethyl)-2,2'-bipyridine]bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl)phenyl]iridium hexafluorophosphate, min. 98%.
SDP
Supplier:  Strem, An Ascensus Company 770340250MG
Encompass
CAS# 2042201-18-1. 250mg. [4,4'-Bis(1,1-dimethylethyl)-2,2'-bipyridine]bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl)phenyl]iridium hexafluorophosphate, min. 98%. . Molecular Weight: 1021.89. Molecular Formula: C40H34F12IrN4P. Color/form: yellow to green yellow solid Strem# 77-0340. http://www.strem.com/catalog/v/77-0340/
Catalog No. NC1692241