Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(2,2-diphenylvinyl)biphenyl 98.0+%, TCI America™
SDP

Catalog No. B27375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B27375G Supplier TCI America Supplier No. B27375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 142289-08-5
Melting Point 208°C
Color Yellow
Molecular Formula C40H30
Quantity 5 g
MDL Number MFCD03093280
Synonym DPVBi
InChI Key UHXOHPVVEHBKKT-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=CC2=CC=C(C=C2)C3=CC=C(C=C3)C=C(C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6
IUPAC Name 1-(2,2-diphenylethenyl)-4-[4-(2,2-diphenylethenyl)phenyl]benzene
Molecular Weight (g/mol) 510.68
PubChem CID 15475895
Formula Weight 510.68
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(2,2-diphenylvinyl)biphenyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.