Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(9H-carbazol-9-yl)-2,2'-dimethylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. B4910200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4910200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4910200MG Supplier TCI America Supplier No. B4910200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 604785-54-8
Molecular Formula C38H28N2
Molecular Weight (g/mol) 512.66
MDL Number MFCD12022459
InChI Key LTUJKAYZIMMJEP-UHFFFAOYSA-N
Synonym 4,4′C-Di(9H-carbazol-9-yl)-2,2′C-dimethyl-1,1′C-biphenyl, CDBP
PubChem CID 22020378
IUPAC Name 9-[4'-(9H-carbazol-9-yl)-2,2'-dimethyl-[1,1'-biphenyl]-4-yl]-9H-carbazole
SMILES CC1=C(C=CC(=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)C1=C(C)C=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 512.66
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(9H-carbazol-9-yl)-2,2′-dimethylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.