Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(chloromethyl)-2,2'-bipyridyl 98.0+%, TCI America™
SDP

Catalog No. B35721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35721G Supplier TCI America Supplier No. B35721G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity building block for synthesis, suitable for use in catalysis and metal complex development.

Chemical Identifiers

CAS 138219-98-4
Molecular Formula C12H10Cl2N2
Molecular Weight (g/mol) 253.126
InChI Key VZKSOWFFOHQARA-UHFFFAOYSA-N
PubChem CID 10562811
IUPAC Name 4-(chloromethyl)-2-[4-(chloromethyl)pyridin-2-yl]pyridine
SMILES C1=CN=C(C=C1CCl)C2=NC=CC(=C2)CCl

Specifications

Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 253.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(chloromethyl)-2,2′-bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.